Tlačiť
Quick release tubing connectors, straight
Dodávateľ: KARTELL
70434 70435 70436 70536 70535
229-0700EA 23.1 EUR
229-0700 229-0701 229-0702 229-0730 229-0729
Quick release tubing connectors, straight
Fitinky a konektory
LDPE, transparent, unbreakable.
  • Good chemical resistance
  • Good thermal resistance (operating temperature up to 80 °C)
  • Unbreakable, provides a very good seal
With tapered ends. Two connecting parts. Provides an excellent seal, but can be released easily at any time. Not suitable for partial vacuums.