Tlačiť
Sample loops for Rheodyne® valves
Dodávateľ: VICI JOUR RESEARCH
JR-SLR-111 JR-SLR-112 JR-SLR-112-77 JR-SLR-113 JR-SLR-113-77 JR-SLR-114 JR-SLR-114-77 JR-SLR-115 JR-SLR-115-77 JR-SLR-116 JR-SLR-116-77 JR-SLR-117 JR-SLR-117-77 JR-SLR-118 JR-SLR-118-77 JR-SLR-111-77
554-1030EA 88.25 EUR
554-1030 554-1031 JOURJR-SLR-112-77 554-1032 JOURJR-SLR-113-77 JOURJR-SLR-114 JOURJR-SLR-114-77 554-1033 JOURJR-SLR-115-77 JOURJR-SLR-116 JOURJR-SLR-116-77 554-1034 JOURJR-SLR-117-77 554-1035 JOURJR-SLR-118-77 JOURJR-SLR-111-77
Sample loops for Rheodyne® valves
Chromatografické potřeby Chromatography Sample Loops
These stainless steel 316 sample loops are cut using an electrolytic cutting process followed by ultrasonic and high pressure steam cleaning to remove contaminants, dried and capped with softener free caps to avoid the recontamination of the tubing.
For valves 1/16" O-Ø.