Tlačiť
Back pressure regulator
Dodávateľ: VICI JOUR RESEARCH
JR-BPR2 JR-BPR1 JR-BPR3
JOURJR-BPR2EA 225.03 EUR
JOURJR-BPR2 554-1492 JOURJR-BPR3
Back pressure regulator
Ventily Ventily nevratné
This regulator installs on the detector drain line and prevents bubble formation in the flow cell during outgassing. It has an adjustable back pressure from 0 to 20 bar (15 to 300 psi) without disconnecting from the system. Other sizes available.
  • Improves baseline stability
  • Low dead volume
  • Biocompatible