Tlačiť
Nuts with double ferrules
Dodávateľ: VICI JOUR RESEARCH
JR-M5004 JR-5004-5 JR-5510-5 JR-5580-5 JR-5511-5
554-1067EA 0 EUR
554-1067 JOURJR-5004-5 JOURJR-5510-5 JOURJR-5580-5 JOURJR-5511-5
Nuts with double ferrules
Fitinky a konektory
These VICI Jour® nuts incorporate a unique PEEK double ferrule which grips the tubing in two places for added security, especially with PEEK or Tefzel tubing. For virtually any ¹/₁₆'' OD fittings eg. Parker, Swagelok, Waters, Valco, Rheodyne, Upchurch.
  • Inert all-PEEK construction
  • Fingertight and hex-head versions
  • Rated up to 420 bar (6000 psi)