Tlačiť
Overalls, Paint-tex® plus, PP and SMS
Dodávateľ: ZVG BADEN WUERTEMBERG
30763-00 30764-00 30765-00
114-3421EA 24.35 EUR
114-3421 114-3422 114-3423
Overalls, Paint-tex® plus, PP and SMS
Jednorázové oděvy
These overalls are tough, three ply, and non linting. They are made of PP with a layer of breathable SMS in between.
  • Anti-static
  • High abrasion resistance
  • Two-way zip closure with protective flap
  • Elasticated at the hood, waist, wrists and ankles
Colour: Light grey
Certifikácie: Cat. III, type 5/6