Tlačiť
Cleanroom face masks, Kimtech™ M3, PE/BiCoSof™
Dodávateľ: KIMBERLY CLARK
62484 62465 62466
111-0151EA 255.52 EUR
111-0151 111-0050 113-0344
Cleanroom face masks, Kimtech™ M3, PE/BiCoSof™
Masky Roušky pro čisté prostory
The pleat-style masks have soft ties and a fully enclosed bendable nosepiece.
  • Outer layer of porous PE film protects from contamination
  • Excellent breathability with filtration of ≥99% at 0,5 µm
  • Inner facing made of white BiCoSof™ and fully enclosed, soft, malleable aluminium nosepiece
Suitable for Class 10, ISO 4, M2.5 cleanrooms.
Balenie: 50 per bag, 6 or 10 bags per case.