Tlačiť
Filling and venting closures, Nalgene®
Dodávateľ: Thermo Fisher Scientific
2162-0830 2162-0831 2162-0531
216-8912EA 73 EUR
216-8912 216-8913 216-8911
Filling and venting closures, Nalgene®
Uzávěry Uzávěry k láhvím
PP, white with TPE seals and TPE caps.
  • Fit all large Nalgene® carboys or bottles with 53 mm (53 B) or 83 mm (83 B) screw caps
  • Choice of 6,3 mm or 12,7 mm tube for the 83 mm cap (83 B)
  • Autoclavable
Screw closures for the sterile liquid transfer of nutrient media, biological reagents, pure water and chemicals. For applications requiring the use of bioreactors and fermenters.
Informace o dodávce: Pack includes the cap, Nalgene® platinum-treated silicone tubing for aspirating and dispensing and a splash guard for filling.