Tlačiť
Joint clamps for conical ground joints
Dodávateľ: witeg Labortechnik
0.131.310 0.131.312 0.131.314 0.131.319 0.131.324 0.131.329 0.131.345
201-7101EA 4.81 EUR
201-7101 201-7102 201-7103 201-7104 201-7105 201-7106 201-7109
Joint clamps for conical ground joints
Svorky Svorky pro kónické NZ
POM, coloured. For firm, securely fitting conical ground joints. These clamps are made of plastic to avoid scratching the glass, which can happen with conventional metal joint clamps.
  • Very good chemical resistance
  • Heat resistant up to +150 °C
  • Sturdy, easy to install and remove