Tlačiť
Clamps
Dodávateľ: USBECK LABORGERATE
2540 2550 2560 2570
241-7314EA 11.31 EUR
241-7314 241-7315 241-7316 241-0254
Clamps
Svorky Svorky pro běžné účely
Zinc die-cast, powder coated.
  • The jaws of the clamps are covered with layer of cork to protect the clamped items
  • Without bosshead for fixing to stand
  • With wing screws for rapid easy clamping