Tlačiť
Cell scrapers, Falcon®
Dodávateľ: Corning
353085 353086 353087 353089
734-0385EA 255.11 EUR
734-0385 734-0386 734-0387 734-1111
Cell scrapers, Falcon®
Škrabky buněčné
PS handle, TPE blade. Designed to provide maximum accessibility to the growth surfaces of a variety of culture vessels.
  • Cross-ribbed PS handle provides greater rigidity, to ensure better control while scraping cells
  • Highly compliant TPE blade pivots to provide multiple angles to remove cells from the entire growth surface
Objednávacie informácie: Supplied individually wrapped in peel-open packages.