Tlačiť
Transport carriers, Nalgene®
Dodávateľ: Thermo Fisher Scientific
7130-0010 7134-0010
139-3110EA 56.16 EUR
139-3110 215-8118
Transport carriers, Nalgene®
Přepravky Boxy přepravní
These bottle carriers allow for safe transport and storage of equipment, such as bottles, and containers. They include a partition wall and an integral carrying handle.
  • Good chemical resistance
  • Heat resistant up to +120 °C
  • Rigid, with two liquid tight compartments
Colour: white.