Tlačiť
Burettes, lateral stopcock, Schellbach
Dodávateľ: HIRSCHMANN
3240160 3240170 3240175 3240360 3240370 3240375
612-9967EA 125.38 EUR
612-9967 612-9968 612-9969 612-9970 612-9971 612-9972
Burettes, lateral stopcock, Schellbach
Byrety
DURAN®, borosilicate glass, class AS.
  • Calibrated to deliver 'Ex', conformity certified (DE-M marking) with dated batch identification
  • Schellbach stripe, ring marks at major graduations
  • Blue graduations
Scale length: 500 mm
Scale length: 500 mm
Certifikácie: DIN EN ISO 385