Tlačiť
Burettes, lateral stopcock, Schellbach, SILBERBRAND
Dodávateľ: BRAND
13563 13566 13568
612-9964EA 131.57 EUR
612-9964 612-9965 612-9966
Burettes, lateral stopcock, Schellbach, SILBERBRAND
Byrety
Borosilicate glass 3.3, class B.
  • Calibrated to deliver 'TD, Ex'
  • Schellbach stripe, short marks at major graduations
Length: 800 mm
Length: 800 mm
Certifikácie: DIN EN ISO 385