Tlačiť
Burettes, straight stopcock, Schellbach, BLAUBRAND®
Dodávateľ: BRAND
12484 12487 12488
612-9958EA 139.19 EUR
612-9958 612-9959 612-9960
Burettes, straight stopcock, Schellbach, BLAUBRAND®
Byrety
Borosilicate glass 3.3, class AS, DE-M marking.
  • Calibrated to deliver 'TD, Ex'
  • Schellbach stripe, ring marks at major graduations
  • Marks and inscriptions in high contrast blue enamel represent the optimum combination of resistance and readability
Length: 800 mm
DIN EN ISO 385
Objednávacie informácie: Supplied with a batch certificate. Burettes are also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate.