Tlačiť
Kryoboxy, barevné, Nalgene®
Dodávateľ: Thermo Fisher Scientific
867013-0240 867013-0241 867013-0242 867013-0243 867013-0244 867013-0245
479-0057EA 44.56 EUR
479-0057 479-0058 479-0059 479-0060 479-0061 479-0062
Kryoboxy, barevné, Nalgene®
Krabice Kryoboxy
PC, autoclavable.
For storage of cryovials from –196 to +121 °C. Colours match cryogenic vial colour coders.
Transparent lid with grid allows easy viewing of contents.
Set: assortment with 4 boxes of each colour (red, yellow, green, blue, grey and white).