Tlačiť
Labtop coolers, Nalgene®
Dodávateľ: Thermo Fisher Scientific
5115-0012 5115-0032 5116-1300 5116-1600 DS5116-0012 DS5116-0032
471-0000EA 147.81 EUR
471-0000 471-0001 471-0002 471-0003 479-3460 479-3461
Labtop coolers, Nalgene®
Krabice Chladící/Mrazící boxy/Sáčky
Labtop coolers −20 °C.
PC, blue.
Lower section filled with a non toxic gel, with handle.
471-0000/471-0001: Will maintain a temperature below –15 °C for up to 2 hours.
Sixteen inserts included convert 1,5 ml holes to accept either 0,5 or 0,2 ml tubes.
Labtop coolers 0 °C.
PC, green.
Lower section filled with a non toxic gel, with handle.
479-3461: Will maintain a temperature below 1 °C for up to 5 hours, gel-filled white lid.
479-3460: Will maintain a temperature below 1 °C for up to 3,5 hours, non-filled clear lid.
471-0002/-0003: Will maintain a temperature below 1 °C for up to 3,5 hours, non-filled clear lid.
Sixteen inserts included convert 1,5 ml holes to accept either 0,5 or 0,2 ml tubes.
471-0001: Will maintain a temperature below –15 °C for up to 2 hours.
Sixteen inserts included convert 1,5 ml holes to accept either 0,5 or 0,2 ml tubes.