Tlačiť
Bottles, wide neck, with screw cap, Nalgene®
Dodávateľ: Thermo Fisher Scientific
2100-0004 2100-0008 2100-0016 2100-0032
215-7101EA 127.96 EUR
215-7101 215-7102 215-7103 215-7104
Bottles, wide neck, with screw cap, Nalgene®
Láhve Láhve plastové
FEP, translucent, with ETFE screw cap.
  • Maximum chemical resistance
  • Temperature resistant from −105 to +150 °C, leakproof
  • Autoclavable
These bottles have the highest chemical and corrosion resistance. Suitable for applications at low and high temperatures, trace analyses, and for many types of work with organic solvents.