Tlačiť
Packaging bottles, wide neck, with screw cap, Nalgene®
Dodávateľ: Thermo Fisher Scientific
312189-0001 312189-0002 312189-0004 312189-0032 312185-0001 312185-0002 312189-0008 312185-0008 312185-0016 312189-0016 312185-0004 312185-0032
215-1138EA 1152.83 EUR
215-1138 215-1139 215-1140 215-1162 215-1448 215-1449 215-1459 215-1451 215-1452 215-1472 215-1450 215-1458
Packaging bottles, wide neck, with screw cap, Nalgene®
Láhve Láhve plastové
HDPE, translucent or amber with PP screw cap.
  • Good chemical resistance
  • Heat resistant from −100 to +120 °C
  • Leakproof
Economical breakproof bottles for storing and transporting powders or pastes. Bottles coloured amber to protect light-sensitive substances.