Tlačiť
Bottles, wide neck, with screw cap, Nalgene®
Dodávateľ: Thermo Fisher Scientific
2107-0008 2107-0016 2107-0032
215-7244EA 93.86 EUR
215-7244 215-7245 215-7246
Bottles, wide neck, with screw cap, Nalgene®
Láhve Láhve plastové
Polymethylpentene PMP, transparent with PP screw cap.
  • High level of chemical resistance
  • Temperature resistant up to +145 °C, autoclavable
  • Leakproof
Can be used anywhere in the laboratory and for sample collection. Wide opening for rapid filling and dispensing of powders, pastes and liquids. Good clarity for viewing bottle contents.