Tlačiť
Wash bottles, narrow neck, autoclavable, Nalgene®
Dodávateľ: Thermo Fisher Scientific
2405-0500 2405-1000
215-8108EA 112.72 EUR
215-8108 215-8107
Wash bottles, narrow neck, autoclavable, Nalgene®
Láhve Střičky
PPCO, translucent, PP spray cap.
  • Very good chemical resistance, heat resistant up to +121 °C
  • Flexible, leakproof closure
  • With blue 'Autoclavable' label and printed size code