Tlačiť
Wash bottles, wide neck with coloured caps, Azlon®
Dodávateľ: DWK LIFE SCIENCES
WGW233P WGW237P WGW239P WGW235P WGW230P WGW236P WGW232P WGW234P WGW238P WGW231P WGW443P WGW444P WGW445P WGW446P WGW447P WGW229P
215-0070EA 33.01 EUR
215-0070 215-0072 215-0074 215-0075 215-0596 215-0598 215-0069 215-0071 215-0073 215-0068 215-0601 215-0602 215-0603 215-0604 215-0605 AZLOWGW229P
Wash bottles, wide neck with coloured caps, Azlon®
Láhve Střičky
LDPE bottle with PP spray cap.
  • Closures feature precision jet tip for fine control
  • Colour coded for easy identification
  • Available in three sizes and five colours
Bottles with wide neck for easy filling. Fitted with colour coded closures that maintain their shape in use.