Tlačiť
Safety wash bottles, narrow/wide neck, with printed name
Dodávateľ: VITLAB
133381 135181 135281 135381
215-6764EA 136.78 EUR
215-6764 VITL135181 VITL135281 VITL135381
Safety wash bottles, narrow/wide neck, with printed name
Láhve Střičky
LDPE, translucent with PP screw cap.
  • Good chemical resistance
  • Heat resistant up to 80 °C (up to 90 °C for brief periods)
Coloured inscription of contents in German, English and French.