Tlačiť
Wash bottles, wide neck with coloured caps
Dodávateľ: KARTELL
1637-00 1638-00 1637-04 1638-04 1637-06 1638-06 1637-10 1638-10
215-9010EA 62.89 EUR
215-9010 215-9011 215-9012 215-9013 215-9014 215-9015 215-9016 215-9017
Wash bottles, wide neck with coloured caps
Láhve Střičky
LDPE, translucent.
  • Good chemical resistance
  • Temperature resistant up to 75 °C
  • Colour coded spray caps in four colours
Wide neck for easy, safe filling. Soft, easily squeezable with moulded graduation. Leakproof since cap and outlet tube made from one piece. Colour coded spray caps.