Tlačiť
Cell culture roller bottles, DURAN®
Dodávateľ: DWK LIFE SCIENCES
217726856 217728651
391-0670EA 341.76 EUR
391-0670 391-0672
Cell culture roller bottles, DURAN®
Láhve Láhve pro rolery
DURAN®, borosilicate glass 3.3, clear, with DIN thread (GL 45).
Supplied with blue PP screw cap and pouring ring.