Tlačiť
Caps for narrow neck aluminium bottles
Dodávateľ: TOURNAIRE-EMBALLAGE
1026 1659 1664
TOUR1026EA 1.6 EUR
TOUR1026 216-5257 216-5258
Caps for narrow neck aluminium bottles
Uzávěry Uzávěry k láhvím
PP or PE, white.
  • Only suitable for 50 – 4000 ml bottles
  • Aluthene insert ensures absolute leakproof properties, even with aggressive media
  • Tamper-evident cap for safety and control