Tlačiť
Economy bottles, narrow neck, round, HDPE, with screw cap, Nalgene®
Dodávateľ: Thermo Fisher Scientific
2089-0002 2089-0004 2089-0008 2089-0016 2089-0032
215-0998EA 86.64 EUR
215-0998 215-1027 215-1028 215-1029 215-1030
Economy bottles, narrow neck, round, HDPE, with screw cap, Nalgene®
Láhve Láhve plastové
HDPE, translucent with PP screw cap.
  • Good chemical resistance
  • Heat resistant from –100 to +120 °C
  • Leakproof
Narrow neck economy bottles are lightweight versions of the original Nalgene® Lab Quality Bottle, suitable for lighter-duty, general purpose applications.