Tlačiť
Culture media bottles, square, sterile, Nalgene®
Dodávateľ: Thermo Fisher Scientific
342020-0060 342020-0125 342020-0250 342020-1000 342020-2000 342020-9500 342020-0500 342020-0030 342024-0125 342024-0250 342024-0030 342024-0060 342024-0500 342024-1000
216-0307EA 542.32 EUR
216-0307 216-0308 216-0309 216-0311 216-0312 216-0609 216-0310 216-0306 216-0381 216-0382 216-0379 216-0380 216-0383 216-0384
Culture media bottles, square, sterile, Nalgene®
Láhve Láhve na média
PETG.
  • Leakproof closure, break-resistant and durable
  • Transparency and reduced permeability to carbon dioxide and oxygen allow media storage up to 6 months
  • Bottles and closures are radiation sterilised and non-pyrogenic
An inexpensive alternative to glass bottles for storage and transport of media. Available with or without a natural HDPE screw cap closure. The caps for the 30 to 1000 ml bottles are a natural off-white colour. The cap for the 2000 ml bottle is white. Bottles and closures are not autoclavable. Packaged in shrink-wrapped trays.