Tlačiť
Culture media bottles, square, sterile, Nalgene®
Dodávateľ: Thermo Fisher Scientific
342044-1000 342040-0250 842040-0650 842040-1000 842040-0125
215-1851EA 203.77 EUR
215-1851 392-0378 392-0381 392-0397 NALG842040-0125
Culture media bottles, square, sterile, Nalgene®
Láhve Láhve na média
PET, clear, with thread or complete with white HDPE screw caps.
  • Good chemical resistance
  • Temperature resistant from −40 to +70 °C
  • Breakproof, suitable for airfreight, leakproof closure
Light breakproof bottles with outstanding gas diffusion properties for storing and transporting liquid media, buffer solutions and sera. Bottles and caps are gamma-sterilised and pyrogen-free. They are non-cytotoxic. With textured graduations and inner sealing ring for use of tamper-evident caps.
Certifikácie: Complying with the requirements of USP, Class VI, USP 661 and European Pharmacopeia for Abnormal Toxicity Standards.
Balenie: Packed in shrink-wrapped layers. Layers are double-packed for use in critical environments.