Tlačiť
Honey jars
Dodávateľ: MUELLER KREMPEL
14.027.21 14.027.22 14.027.64
216-3605EA 27.04 EUR
216-3605 216-3606 MUEL14.027.64
Honey jars
Nádoby Nádoby víceúčelové
Glass, clear, without lid.
  • Good chemical and physical properties
  • For use in applications without high thermal loads
  • Metal screw cap available separately (see 216-3664 and 216-3664)
Perfect for sample collection, storage and transport of powders, pastes and liquids.