Tlačiť
Plynové mycí láhve, PFA, PTFE frity a spojky trubek, BOLA
Dodávateľ: Bohlender
A118-01 A118-02 A118-03
201-0015EA 417.57 EUR
201-0015 201-0016 201-0017
Plynové mycí láhve, PFA, PTFE frity a spojky trubek, BOLA
Láhve Promývačka plynů
Transparent PFA bottle with white PTFE frit.
  • Pore-free, resistant to practically all chemicals
  • Fine-pore frit of 3 µm ensures optimal pearling of the gas to be washed
  • Temperature resistance from −200 to +250 °C
Stable, robust bottles.