Tlačiť
Dropping bottles, LDPE, Nalgene®
Dodávateľ: Thermo Fisher Scientific
2752-9025 2752-9050 2752-9125
215-1061EA 70.5 EUR
215-1061 215-1062 215-1063
Dropping bottles, LDPE, Nalgene®
Láhve Lahvičky kapací
LDPE, transparent with colour-coded (red, green, blue, white and yellow) PP screw caps and LDPE dosing tip.
  • Good chemical resistance, heat resistant up to 80 °C
  • Drop control tip snaps securely into place, drops are dispensed individually
  • Flexible for easy handling, leakproof
  • Delivers 40 µl drops (based on water; viscosity affects drop size)
Reagent identification is very easy with the varied colours of the screw caps. These dropping bottles provide reliable, repeatable dispensing of reagents and other liquids. They serve as a very good alternative to pipettes or other conventional dispensing devices. They are suitable for most biotech, diagnostic and pharmaceutical applications.