Tlačiť
Centrifuge bottles, PPCO/PC/PS, with screw cap, Nalgene®
Dodávateľ: Thermo Fisher Scientific
3143-0175 3144-0175 3145-0175
525-2431EA 163.26 EUR
525-2431 525-2441 525-2451
Centrifuge bottles, PPCO/PC/PS, with screw cap, Nalgene®
Láhve Lahvičky odstřeďovací
Conical bottom and rimless, graduated.
  • Withstands forces up to 27 500×g and 5800×g for 525-2451
  • Suitable for use in refrigerated or non refrigerated centrifuges
  • Must be used with adapter for conical-bottom centrifuge bottles 525-2400