Tlačiť
Demižóny, široké hrdlo, PP nebo PC, se šroubovacím uzávěrem a hrdlem, Nalgene®
Dodávateľ: Thermo Fisher Scientific
2317-0020 2317-0050 2319-0020 2319-0050 2319-0130
216-8791EA 319.7 EUR
216-8791 216-8792 216-8771 216-8772 216-4411
Demižóny, široké hrdlo, PP nebo PC, se šroubovacím uzávěrem a hrdlem, Nalgene®
Láhve Demižóny
PP, translucent with white PP screw cap and PP spigot with TPE seal, or PC, clear with white PP screw cap and PP spigot with TPE seal.
  • PP: Very good chemical resistance, ideal for storing, transporting and dispensing solutions, culture media and distilled water/PC: Good chemical resistance, ideal for storing, transporting and dispensing sterile water, standard reagents and culture media
  • PP: Temperature resistant from –10 to +135 °C, autoclavable/PC: Temperature resistant from –100 to +135 °C, autoclavable
  • Leakproof closure
Wide mouth opening for easy filling and cleaning. For better durability and leak resistance the drain cock thread is moulded in one piece with the carboy, not welded on. Graduated.