Tlačiť
Aspirator bottles, glass, PYREX®
Dodávateľ: DWK LIFE SCIENCES
1541/18D 1541/16D 1541/22D
215-0353EA 410.75 EUR
215-0353 215-0352 215-0354
Aspirator bottles, glass, PYREX®
Láhve Aspirátory
Made of PYREX®, borosilicate glass.
  • Excellent chemical and thermal resistance
  • White labelling area
  • Unground neck to accept rubber stoppers
With NS 29/32 ground side arm socket to accept spigot assemblies.
Certifikácie: ISO 4796
Objednávacie informácie: Spigot for side arm socket (size NS 29/32) should be ordered separately.