Tlačiť
Spray bottles, LaboPlast®
Dodávateľ: BURKLE
0309-1100 0309-1025 0309-1050
215-6513EA 8.26 EUR
215-6513 215-6510 215-6511
Spray bottles, LaboPlast®
Láhve Lahvičky s rozprašovačem
LDPE, translucent, PP spray attachment.
  • Good chemical resistance, good temperature resistance
  • Spray nozzle adjustment can be varied by rotation
  • Pressureless spray bottle
  • Media-contacting parts PP, PE and stainless steel
Spray, choice of fine mist or focused jet spray functions to disperse fluids.