Tlačiť
Measuring cylinders, conical, PMP
Dodávateľ: KARTELL
1422 1423 1424 1425
213-1821EA 17.61 EUR
213-1821 213-1822 213-1823 213-1824
Measuring cylinders, conical, PMP
Válce Válce odměrné
PMP (TPX), crystal clear.
  • Very good chemical resistance and thermal resistance up to 170 °C
  • Robust, autoclavable
  • Liquid volume is easy to read
Ideal for a wide range of applications. With moulded graduations. Broad base for stability.