Tlačiť
Measuring beakers with handle
Dodávateľ: VITLAB
440941 442941 443941 445941 446941 447941 441941 444941
213-1611EA 3.94 EUR
213-1611 213-1613 213-1614 213-0257 213-0255 213-0256 213-0262 213-0263
Measuring beakers with handle
Kádinky
PP, highly translucent.
  • Good chemical resistance
  • Resistant to heat up to 121 °C (for 20 minutes), prolonged use 100 °C
  • Smooth surfaces for easy, fast cleaning
Ideal for general laboratory use and a wide variety of other applications. With embossed scale and spout.