Tlačiť
Measuring beakers with handle
Dodávateľ: MORGAN ADVANCED MATERIALS HALD
L51/3A L51/8 L51/10
213-5105EA 155.02 EUR
213-5105 213-5115 213-5120
Measuring beakers with handle
Kádinky
Porcelain, glazed white except for the base.
  • Exceptional chemical resistance
  • Very good thermal resistance
  • Robust, heavy construction
Cylindrical, with spout. With very easy-to-read, internal graduations. Label with the nominal volume on the outside of the measuring beaker.