Tlačiť
Beakers, low form
Dodávateľ: VITLAB
110205 110305 110405 110605 110905 111005
213-1602EA 29.16 EUR
213-1602 213-1603 213-1604 213-1605 213-1606 213-1609
Beakers, low form
Kádinky
PFA, translucent, with spout.
  • Excellent chemical resistance
  • High thermal resistance from –270 to +250 °C
  • Autoclavable
Ideal for demanding applications. Not for use on hotplates. With engraved graduation.
ISO 7056
Certifikácie: ISO 7056