Tlačiť
Autoclaving baskets, Nalgene®
Dodávateľ: Thermo Fisher Scientific
6917-0127 6917-0150 6917-0230
489-2150EA 44.86 EUR
489-2150 489-2151 489-2152
Autoclaving baskets, Nalgene®
Koše a košíky Košíky autoklávovatelné
PP.
  • Smooth surfaces will not scratch plastic or glass
  • Provides airflow for venting and drying
  • Three sizes available
  • Nestable for storage
These autoclavable baskets are designed for holding labware during autoclaving and washing.
Colour: White