Tlačiť
Biohazard bag holders, Clavies®, SP Bel-Art
Dodávateľ: Bel-Art Products, a Part of SP
F131920001 F131920002 F131920003 F131920102
129-1026EA 112.72 EUR
129-1026 129-1012 129-1027 BELAF131920102
Biohazard bag holders, Clavies®, SP Bel-Art
Stojany Držáky sáčků
To keep biohazard disposal bags open and ready to use.
  • Stainless steel wire frame
  • Base in orange PP, which serves as a leakage tray
  • Autoclavable at 121 °C to maintain sterility
Informace o dodávce: Delivered unassembled.