Tlačiť
Adapters, delivery, angled
Dodávateľ: LENZ LABORGLAS
05260001 05260002 05260003 05260004 05263001 05263002 05263003 05263004
271-2401EA 24.55 EUR
271-2401 271-2402 201-0364 201-3108 201-0365 201-0366 201-0367 271-2403
Adapters, delivery, angled
Komponenty pro destilaci Adaptéry destilační
DURAN®, borosilicate 3.3 glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Standard ground joint socket and delivery tube at an angle of 105°. Available with long and short delivery tubes.