Tlačiť
Adapters, receiver, vacuum, straight or angled
Dodávateľ: LENZ LABORGLAS
05272001 05272002 05272003 05272004 05275001 05275002 05275003 05275004
201-2411EA 30.61 EUR
201-2411 201-0368 201-0369 201-2412 201-2416 201-0370 201-0371 201-2417
Adapters, receiver, vacuum, straight or angled
Komponenty pro destilaci Adaptéry destilační
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Standard ground joint cone and socket with drip point and glass vacuum tubing connector. Available in straight or 105° angled versions.