Tlačiť
Adapters, swan neck, Quickfit®
Dodávateľ: DWK LIFE SCIENCES
MA6/22 MA6/33
201-8576EA 73 EUR
201-8576 201-3010
Adapters, swan neck, Quickfit®
Komponenty pro destilaci Adaptéry destilační
PYREX®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With screwthread joint (size 13) to accept thermometers, air leak tubes etc.