Tlačiť
Bottles, wide neck, with screw cap
Dodávateľ: VITLAB
109497 109597 109697 109797
215-4593EA 169.68 EUR
215-4593 215-4594 215-4595 215-1961
Bottles, wide neck, with screw cap
Láhve Láhve plastové
PFA, translucent, with PFA screw cap.
  • Maximum chemical resistance
  • Temperature resistant from –200 to +250 °C, autoclavable
  • Leakproof
Bottles with high resistance to extreme temperatures, chemicals and corrosion. Inert to practically all chemicals with the exception of molten alkaline metals, fluorine compounds and complex halogen compounds at high temperatures and under pressure. Ideal for liquids, pastes, powders and granular material. Wide mouth bottles with large opening for easy filling and cleaning.