Tlačiť

Florisil® for chromatography, Supelco®

Dodávateľ: Merck

Synonymá: FlorisilMagnesium silicate, aktivní

M-Clarity™ quality level = MQ100

SIAL52462-1KT-FEA 0 EUR
SIAL52462-1KT-F
Florisil® for chromatography, Supelco®
Magnesium silicate

Pozor: Florisil is a registered trademark of U.S. Silica Company

Vzorec: MgO₃Si
Mw: 100,38 g/mol
Číslo MDL: MFCD00078249
Číslo CAS: 1343-88-0
EINECS: 215-681-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR