Tlačiť

Florisil® for chromatography, Supelco®

Dodávateľ: Merck

Synonymá: FlorisilMagnesium silicate, aktivní

M-Clarity™ quality level = MQ100

46384-500G-FEA 216.44 EUR
46384-500G-F
Florisil® for chromatography, Supelco®
Magnesium silicate
Vzorec: MgO₃Si
Mw: 100,38 g/mol
Číslo MDL: MFCD00078249
Číslo CAS: 1343-88-0
EINECS: 215-681-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR