Tlačiť
Centrifuge Tubes, Conical Bottom, Graduated, ASTM
Dodávateľ: GLASSCO
088.202.01 088.202.02 088.202.03_U 088.202.05 088.202.03
GLAS088.202.01EA 131.57 EUR
GLAS088.202.01 GLAS088.202.02 GLAS088.202.03_U GLAS088.202.05 GLAS088.202.03
Centrifuge Tubes, Conical Bottom, Graduated, ASTM
Zkumavky Zkumavky centrifugační
ASTM Graduated centrifuge tubes, with conical bottom.
  • Made from ASTM E-438 Type-1 Class A, Borosilicate 3.3 glass
  • Uniform wall thickness
  • Strudy lip