Tlačiť

Anilin 99,5, AnalaR NORMAPUR® zA

Dodávateľ: VWR Chemicals
Nebezpečí

Synonymá: Aminobenzene, Phenylamine

21425.290EA 155.82 EUR
21425.290 21425.230
Anilin 99,5, AnalaR NORMAPUR® zA
Anilin

* material may darken during storage with no loss in purity

Vzorec: C₆H₅NH₂
Mw: 93.13
Teplota varu: 184 °C (1013 hPa)
Teplota tavenia: –6,2 °C
Hustota: 1.019 g/cm³ (20 °C)
Teplota vzplanutia: 76 °C
Skladovacia teplota: Ambient
Číslo MDL: MFCD00007629
Číslo CAS: 62-53-3
EINECS: 200-539-3
OSN: 1547
ADR: 6.1 II
REACH: 01-2119451454-41
Merck Index: 13,00661

Order Now

Výsledky špecifikačného testu

Appearance colorless to dark-brownish / yellow liquid
Assay (GC) min. 99.5%
Water max. 0.2%
Residue after ignition max. 0.005%
Hydrocarbons Passes Test
Nitrobenzene (C6H5NO2) max. 0.003%
Copper (Cu) max. 0.5 ppm
Iron (Fe) max. 1 ppm
Lead (Pb) max. 1 ppm
Density (20°C) 1.021 - 1.023


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR