Tlačiť
Anti-IgG Goat Polyclonal Antibody (iFluor® 860)
Dodávateľ: AAT BIOQUEST
48016 48017
AATB48016EA 148.01 EUR
AATB48016 AATB48017
Anti-IgG Goat Polyclonal Antibody (iFluor® 860)
Protilátky
AAT Bioquest's iFluor® dyes are optimised for labeling proteins, in particular, antibodies.
  • Purified and quality-tested for consistent results
  • Suitable for multiple applications
  • Purified through affinity chromatography
iFluor® 860 goat anti-mouse IgG (H+L) conjugates are ideal for various NIR imaging applications, including Western blotting, ELISA, protein arrays, tissue section imaging, and in vivo imaging.
Pozor: Research use only.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: iFluor® 860
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse